Order Entry
Mexico
ContactUsLinkComponent
Sulphuric acid ( 0.51 mol/l) in aqueous solution 0.5 N BAKER ANALYZED®
Sulphuric acid ( 0.51 mol/l) in aqueous solution 0.5 N BAKER ANALYZED®
Catalog # JT5640-2
Supplier:  Avantor
CAS Number:  
MOC:  Non-Regulated, With InformationCertificates
undefined
Sulphuric acid ( 0.51 mol/l) in aqueous solution 0.5 N BAKER ANALYZED®
Catalog # JT5640-2
Supplier:  Avantor
Supplier Number:  5640-02
CAS Number:  
MOC:  Non-Regulated, With InformationCertificates

Specifications

  • Pack type:
    PE bottle, narrow mouth
  • Size:
    1 L
  • Cat. no.:
    JT5640-2
  • Supplier No.:
    5640-02

Specifications

Specification Test Results

  • Normality:
    0.4995 - 0.5005
  • Chloride (Cl):
    ≤1 ppm
  • Phosphate (PO₄):
    ≤1 ppm
  • Heavy Metals (as Pb):
    ≤0.3 ppm
  • Trace Impurities - Iron (Fe):
    ≤0.5 ppm

Specification Test Results

About this item

 :
 :98.08 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2796
ADR:8,II
 :Management of Change (MOC) category = NI
EINECS:231-639-5
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589