Starch, soluble from potatoes BAKER ANALYZED® ACS
Catalog # JT4006-1
Supplier: AVANTOR PERFORMANCE MATERIALS MX
CAS Number:
MOC: Non-RegulatedCertificates
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Size:500 g
- Cat. no.:JT4006-1
- Supplier No.:4006-01
Specifications
Specification Test Results
- Solubility:Passes Test
- pH of 2% Solution at 25°C:5.0 - 7.0
- Residue after Ignition:≤0.4 %
- Sensitivity:Passes Test
Specification Test Results
About this item
:
:162.14 g/mol
:Management of Change (MOC) category = N
Storage Temperature:Ambient
EINECS:232-686-4
CAS Number:9005-84-9
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00082026