Order Entry
Mexico
ContactUsLinkComponent
KIMAX® Volumetric Flasks, Cassia, Class A, Kimble Chase
KIMAX® Volumetric Flasks, Cassia, Class A, Kimble Chase
Catalog # 29630-000
undefined
KIMAX® Volumetric Flasks, Cassia, Class A, Kimble Chase
Catalog # 29630-000
Supplier Number:  28066-110
Specifications
  • Capacity:
    110 ml
  • Cat. no.:
    29630-000
  • ST Stopper No.:
    13
  • Supplier No.:
    28066-110
Specifications
About this item
Calibrated to contain
For assaying essential oils according to methods of the U.S. Pharmacopoeia and Essential Oil Association of the U.S.A. White ceramic enamel scale. Supplied with a [ST] glass stopper and marking spot.