Order Entry
Mexico
ContactUsLinkComponent
Sodium molybdate(VI) dihydrate ≥99.5% BAKER ANALYZED® ACS
Sodium molybdate(VI) dihydrate ≥99.5% BAKER ANALYZED® ACS
Catalog # JT3764-1
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
Sodium molybdate(VI) dihydrate ≥99.5% BAKER ANALYZED® ACS
Catalog # JT3764-1
Supplier Number:  3764-01
CAS Number:  
MOC:  Non-RegulatedCertificates

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Size:
    500 g
  • Cat. no.:
    JT3764-1
  • Supplier No.:
    3764-01

Specifications

Specification Test Results

  • Appearance:
    Passes Test
  • Assay (Na₂MoO₄·2H₂O):
    99.5 - 103.0 %
  • Insoluble Matter:
    ≤0.005 %
  • pH of 5% Solution at 25°C:
    7.0 - 10.5
  • Chloride (Cl):
    ≤0.005 %
  • Ammonium (NH₄):
    ≤0.001 %
  • Sulfate (SO₄):
    ≤0.015 %
  • Trace Impurities - Iron (Fe):
    ≤0.001 %
  • Phosphate (PO₄):
    ≤5 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm

Specification Test Results

About this item

 :
 :241.95 g/mol
Merck Index:13,08718
Melting Pt:687 °C
Density:3.28 g/cm³ (20 °C)
 :Management of Change (MOC) category = N
Storage Temperature:Ambient
EINECS:231-551-7
CAS Number:10102-40-6
Formula:Na₂MoO₄·2H₂O
MDL Number:MFCD00149170