Order Entry
Mexico
ContactUsLinkComponent
AlphaTec® 09-928 Chemical resistant gloves, Ansell
Catálogo # 32889-998
undefined
AlphaTec® 09-928 Chemical resistant gloves, Ansell
Catálogo # 32889-998
Número de proveedor:  103644
Especificaciones
  • Glove Color:
    Black
  • Glove size:
    L
  • Ambidextrous / hand-specific:
    Hand-specific
  • Disposable / reusable:
    Reusable
  • Description:
    Gauntlet
  • Length:
    45.7 cm (17⅞")
  • Cuff style:
    Gauntlet
  • Finish:
    Smooth
  • Cat. no.:
    32889-998
  • Model:
    9-928
  • Supplier No.:
    213321
Especificaciones
Acerca de este artículo
Gloves are fully coated with reinforced neoprene coating to provide good all-around resistance to abrasions, cuts, and chemicals.
  • Strong seams prevent hand irritation
  • Good resistance to abrasions, cuts, and chemicals
  • Natural fit
  • Great comfort
Interlock knit cotton liner enhances puncture and snag resistance. Curved fingers and wing thumb provide natural fit and greater comfort. Two-piece lining eliminates seams from working areas to prevent hand irritation. Applications extend to refining and petrochemicals; handling oils, acids, caustics, and alcohols; degreasing; plating; and metal fabrication.