Order Entry
Mexico
ContactUsLinkComponent
Sodium acetate, anhydrous ≥99.0%, OmniPur®, Millipore®
Sodium acetate, anhydrous ≥99.0%, OmniPur®, Millipore®
Catalog # EM-7510
CAS Number:  
Sodium acetate, anhydrous ≥99.0%, OmniPur®, Millipore®
Catalog # EM-7510
Supplier Number:  7510-500GM
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Poly bottle
  • Size:
    500 g
  • Environmentally Preferable:
  • Cat. no.:
    EM-7510
  • Supplier No.:
    7510-500GM

Specifications

Specification Test Results

  • A (260 nm, 0.1 M, water):
    0.01AU max
  • A (280 nm, 0.1 M, water):
    0.01AU max
  • Appearance:
    White crystals
  • Assay:
    99.0% min
  • DNase:
    None detected
  • Heavy metals (as Pb):
    0.001% max
  • pH (5%, water):
    7.5-9.2
  • Phosphatase:
    None detected
  • Protease:
    None detected
  • RNase:
    None detected

Specification Test Results

About this item

Synonyms: Acetic acid sodium salt
 :
 :82.03 g/mol
Merck Index:12,08711
Boiling Pt:881.4 °C (1013 hPa)
Boiling Pt:881.4 °C (1013 hPa)
Melting Pt:324 °C
Density:1.528 g/cm³ (20 °C)
Flash Pt:>250 °C (closed cup)
Storage Temperature:Ambient
EINECS:204-823-8
REACH:01-2119485123-42
CAS Number:127-09-3
Formula:H₃CCOONa
MDL Number:MFCD00012459
Flash Pt:≥ 250 °C (closed cup)