2-Propanol ≥99.9% (by GC), OmniSolv®, Supelco®
Supplier: THOMAS SCIENTIFIC INC MX
Certificates
About this item
Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
CAS Number:67-63-0
:
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
:60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:-89 °C
Density:0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
REACH:01-2119457558-25
:GHS02
:GHS07
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (GC):99.9% min
- Capillary ECD responsive substances (as PCNB):2ng/L max
- Color (APHA):10 max
- Filtered through 0.2 µm filter:To pass test
- Fluorescence (as quinine base):250ppt max
- Form:Clear liquid
- Identity (IR-spectrum):Conforms
- Residue after evaporation:1ppm max
- Titrable acid:0.2µeq/g max
- UV Abs. at 204 nm:1.00AU max
- UV Abs. at 205 nm:0.80AU max
- UV Abs. at 210 nm:0.35AU max
- UV Abs. at 220 nm:0.10AU max
- UV Abs. at 230 nm:0.05AU max
- UV Abs. at 240 nm:0.02AU max
- UV Abs. at 260 nm:0.005AU max
- UV Abs. at 300 nm:0.005AU max
- UV Cut-off:204nm max
- Water (H2O):0.05% max
Specification Test Results
Related Information
Certificates
Search by Lot NumberAYN4_Analytical_Chemistry_-_Chromatography EMD Millipore Start and End with Quality Product Portfolio Brochure EMD Millipore Sustainable Products Brochure MilliporeSigma The New M-Bottle for Solvents.pdf VWR® Chemicals in Poly-Coated Glass Bottles
Product Literature
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together