1-Octanesulphonic acid sodium salt ≥98% for HPLC
Supplier: AVANTOR PERFORMANCE MATERIALS MX
MOC: Non-Regulated
About this item
CAS Number:5324-84-5
:
Formula:C₈H₁₇NaO₃S
MDL Number:MFCD00007544
:216.28 g/mol
Melting Pt:? 300 °C
:Management of Change (MOC) category = N
EINECS:226-195-4
:GHS07
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (as C₈H₁₇SO₃Na) (by acidimetry):≥98.0 %
- UV Absorbance of 0.25 M Solution - 250 nm:≤0.050
- UV Absorbance of 0.25 M Solution - 240 nm:≤0.050
- UV Absorbance of 0.25 M Solution - 230 nm:≤0.050
- UV Absorbance of 0.25 M Solution - 220 nm:≤0.060
- UV Absorbance of 0.25 M Solution - 210 nm:≤0.080
- UV Absorbance of 0.25 M Solution - 200 nm:≤0.200
Specification Test Results
Related Information
Certificates
Search by Lot Number2818 Product Specifications
Additional Product Details
Avantor High-Purity Solvents and Reagents Avantor J.T.Baker® LCMS and HPLC Solvents and Reagents Avantor Portfolio of Chemicals
Product Literature
JTBaker 2818 Datasheet
Datasheet
Frequently Bought Together