

Ethanolamine ≥99.0% (by GC, titration analysis)
Catalog # TCA0297-25G
Proveedor: TCI America
Número CAS:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Especificaciones
- Size:25 g
- Cat. no.:TCA0297-25G
- Supplier No.:A0297-25G
Especificaciones
Resultados de prueba de especificación
- Purity/Analysis Method:99.0% (GC,T)
Resultados de prueba de especificación
Acerca de este artículo
:
Índice Merck:13,03762
Punto de ebullición:170 °C
Punto de ebullición:170 °C
Punto de fusión:10 °C
Densidad:1.02 (20 °C)
Punto de inflamación:93 °C
Punto de inflamación:93 °C
Temperatura de almacenamiento:Ambient
EINECS:205-483-3
UN:2491
ADR:8,III
:61.08 g/mol
:GHS05
:GHS07
Número CAS:141-43-5
Fórmula:NH₂CH₂CH₂OH
Número MDL:MFCD00008183