

Pyridine ACS for HPLC, for spectrophotometry
Catalog # BJ332-4
Supplier: HONEYWELL INTERNATIONAL INC MX
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:4 L
- Cat. no.:BJ332-4
- Supplier No.:332-4
Specifications
Specification Test Results
- Amines by Ninhydrin Test:10 ppm max.
- Free Acid:Passes test
- GC Analysis:99.8 % min.
- Refractive Index (20ºC):1.5085 - 1.5109
- Residue:1 mg/L max.
- Water by Karl Fischer Titration:0.01 % max.
Specification Test Results
About this item
:
Merck Index:13,08060
Boiling Pt:115.25 °C
Boiling Pt:115 °C (1013 hPa)
Melting Pt:–41.55 °C
Melting Pt:-44 °C
Density:0.983 g/cm³ (20 °C)
Flash Pt:12.4 %(V)
Flash Pt:17 °C
Storage Temperature:Ambient
EINECS:203-809-9
UN:1282
ADR:3,II
:79.1 g/mol
:GHS02
:GHS07
CAS Number:110-86-1
Formula:C₅H₅N
MDL Number:MFCD00011732