Mercury (II) acetate ≥98% BAKER ANALYZED® ACS
Supplier: Avantor
MOC: Non-Regulated
About this item
CAS Number:1600-27-7
:
Formula:(H₃CCOO)₂Hg
MDL Number:MFCD00012165
:318.68 g/mol
Merck Index:13,05898
Melting Pt:178 - 180 °C
Density:3.27 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:216-491-1
ADR:6.1,II
:Management of Change (MOC) category = N
UN:1629
:GHS06
:GHS08
:GHS09
Specification Test Results
- Assay ((CH₃COO)₂Hg):≥98.0 %
- Insoluble Matter:≤0.01 %
- Residue after Reduction:≤0.020 %
- Chloride (Cl):≤0.005 %
- Nitrate (NO₃):≤0.005 %
- Sulfate (SO₄):≤0.005 %
- Other Heavy Metals (as Pb):≤0.002 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Mercurous Mercury (as Hg):≤0.4 %
Specification Test Results
Related Information
Certificates
Search by Lot Number2584 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Brand Definition Baker Analyzed
Datasheet
Frequently Bought Together