Order Entry
Mexico
Orders LinkContactUsLinkComponent
D-(-)-Mannitol, powder, BAKER ANALYZED® ACS

D-(-)-Mannitol, powder, BAKER ANALYZED® ACS

PE pail, screw top, 12 kg
Catalog # JT2554-7
Supplier #:  2554-07
D-(-)-Mannitol, powder, BAKER ANALYZED® ACS
Catalog # JT2554-7
CAS Number:  69-65-8
MOC:  Non-RegulatedCertificates
D-(-)-Mannitol, powder, BAKER ANALYZED® ACS
D-(-)-Mannitol, powder, BAKER ANALYZED® ACS

PE pail, screw top, 12 kg

CAS Number:  69-65-8
MOC:  Non-Regulated
Price
$4,516.98
Log in for availability
Minimum order quantity is 1
Product Details & Documents

Powder. Lot analysis on label.

Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,05769
  • Boiling Pt:290 - 295 °C (4 hPa)
  • Boiling Pt:290…295 °C (4 hPa)
  • Melting Pt:164 - 169 °C
  • Density:1.49 g/cm³ (20 °C)
  •  :Management of Change (MOC) category = N
  • Melting Pt:164…169 °C
  • Storage Temperature:Ambient
  • EINECS:200-711-8
Specifications

Specification Test Results

  • ACS - Specific Rotation [Å]²⁵ ^D (+):
    23.3 - 24.3 Degree
  • ACS - Insoluble Matter:
    ≤0.01 %
  • ACS - Loss on Drying at 105°C:
    ≤0.05 %
  • ACS - Residue after Ignition:
    ≤0.01 %
  • ACS - Titrable Acid (meq/g):
    ≤0.0008
  • ACS - Reducing Sugars:
    Passes Test
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm

Specifications

  • Pack type:
    PE pail, screw top
  • Size:
    12 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.