Magnesium sulphate ≥99%, powder BAKER ANALYZED®
Catalog # JT2506-1
Supplier: AVANTOR PERFORMANCE MATERIALS MX
CAS Number:
MOC: Non-RegulatedCertificates
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Size:500 g
- Cat. no.:JT2506-1
- Supplier No.:2506-01
Specifications
Specification Test Results
- Assay (MgSO₄) (ignited basis):≥99.0 %
- Loss on Ignition:≤2.0 %
- Chloride (Cl):≤0.001 %
- Nitrate (NO₃):≤0.004 %
- Ammonium (NH₄):≤0.004 %
- Calcium (Ca):≤0.06 %
- Heavy Metals (as Pb):≤0.001 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Manganese (Mn):≤0.001 %
- Trace Impurities - Arsenic (As):≤1 ppm
Specification Test Results
About this item
:
:120.37 g/mol
Merck Index:12,05731
Melting Pt:1127 °C
Density:2.65 g/cm³ (20 °C)
:Management of Change (MOC) category = N
Storage Temperature:Ambient
EINECS:231-298-2
CAS Number:7487-88-9
Formula:MgSO₄
MDL Number:MFCD00011110