Order Entry
Mexico
ContactUsLinkComponent
 
KIMBLE® RAY-SORB® Amber Flat Bottom Short Neck Boiling Flask, DWK Life Sciences
  89001-270
undefined
KIMBLE® RAY-SORB® Amber Flat Bottom Short Neck Boiling Flask, DWK Life Sciences
  89001-270
 :  25057-250

 

  • ST Joint No.:
    24/40
  • Capacity:
    250 ml
  • Cat. no.:
    89001-270
  • O.D.×H:
    83 x 133 mm
  • Supplier No.:
    25057-250

 

 

Full-length, short neck flasks are RAY-SORB® processed to provide protection to solutions sensitive to short-wavelength light.

Designed from ASTM Specification E 1403, Type I, Class 4 requirements.