Order Entry
Mexico
ContactUsLinkComponent
Methanol ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Methanol ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Catalog # EM-MX0486-1
CAS Number:  
undefined
Methanol ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Catalog # EM-MX0486-1
Supplier Number:  MX0486-1
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass bottle
  • Size:
    4 L
  • Environmentally Preferable:
  • Cat. no.:
    EM-MX0486-1
  • Supplier No.:
    MX0486-1

Specifications

Specification Test Results

  • Assay (GC):
    99.9% min
  • Color (APHA):
    10 max
  • Filtered through 0.2 µm filter:
    To pass test
  • Fluorescence as quinine at 254 nm:
    1ppb max
  • Fluorescence as quinine at 365 nm:
    1ppb max
  • Form:
    Clear liquid
  • Gradient at 235 nm:
    1mAU max
  • Gradient at 254 nm:
    1mAU max
  • Identity (IR-spectrum):
    Conforms
  • Metals, suitable for LC/MS:
    To pass test
  • MS purity, ESI(+), largest response as reserpine:
    50ppb max
  • MS purity, ESI(-), largest response, p-nitrophenol:
    50ppb max
  • Residue after evaporation:
    1ppm max
  • Titrable acid:
    0.3µeq/g max
  • Titrable base:
    0.2µeq/g max
  • UV Abs. at 210 nm:
    0.3AU max
  • UV Abs. at 220 nm:
    0.1AU max
  • UV Abs. at 230 nm:
    0.04AU max
  • UV Abs. at 240 nm:
    0.03AU max
  • UV Abs. at 250 nm:
    0.01AU max
  • UV Abs. at 270 nm:
    0.01AU max
  • UV Abs. at 400-280 nm:
    0.005AU max
  • UV Cut-off:
    205nm max
  • Water (H2O):
    0.05% max

Specification Test Results

About this item

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
 :
 :32.04 g/mol
EINECS:200-659-6
UN:1230
REACH:01-2119433307-44
 :GHS02
 :GHS06
 :GHS08
Storage Temperature:Ambient
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595
Boiling Pt:64.6
Melting Pt:-98
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 (closed cup)
ADR:3 II
Merck Index:13,05984