


Methanol ≥99.5%, OmniSolv® for pesticide residue analysis, Supelco®
EM-MX0484-1
:
- Pack type:Glass bottle
- Size:4 L
- Cat. no.:EM-MX0484-1
- Supplier No.:MX0484-1
- Assay (GC):99.5% min
- Aldehydes and ketones:10ppm max
- Cap. ECD responsive substances as PCNB (peak):5ng/L max
- Cap. ECD responsive substances as PCNB (total):50ng/L max
- Cap. FID responsive substances as decane (peak):2µg/L max
- Cap. FID responsive substances as decane (total):20µg/L max
- Color (APHA):10 max
- Filtered through 0.2 µm filter:To pass test
- Form:Clear liquid
- Infrared spectrum:Conforms to standard
- Residue after evaporation:1ppm max
- Titrable acid:0.3µeq/g max
- Water (H2O):0.05% max
:
:32.04 g/mol
:13,05984
:64.6 °C (1013 hPa)
:64.6 °C (1013 hPa)
:–98 °C
:-98 °C
:0.7918 g/cm³ (20 °C)
:11 °C (closed cup)
:Ambient
:200-659-6
:1230
:3,II
:01-2119433307-44
:GHS02
:GHS06
:GHS08
:67-56-1
:CH₃OH
:MFCD00004595