Order Entry
Mexico
ContactUsLinkComponent
Methanol, anhydrous ≥99.9% for synthesis
Methanol, anhydrous ≥99.9% for synthesis
Catalog # BJAS230-1
Product available on GSA Advantage®
CAS Number:  
undefined
Methanol, anhydrous ≥99.9% for synthesis
Catalog # BJAS230-1
Supplier Number:  AS230-1
Product available on GSA Advantage®
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle, septum-sealed
  • Size:
    1 L
  • Cat. no.:
    BJAS230-1
  • Supplier No.:
    AS230-1

Specifications

Specification Test Results

  • Appearance:
    Passes test
  • Color (APHA):
    10 max.
  • GC Analysis:
    99.9 % min.
  • Residue:
    3 mg/L max.
  • Water by Karl Fischer Titration:
    50 ppm max.

Specification Test Results

About this item

 :
 :32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.7 °C
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Melting Pt:-98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C
Flash Pt:11 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
 :GHS02
 :GHS06
 :GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595