STANSOLV® Flock-Lined Nitrile Gloves, Mapa Professionnel
About this item
Offer hand protection for industrial applications such as plating, metal finishing, chemical processing, abrasive material handling, maintenance, and acid etching.
- Curved fingers and tapered wrist for natural, comfortable wear
- Easy donning and doffing
- Flock lining absorbs perspiration for added comfort
- Flat cuffs
- Hand-specific
Double-dipped gloves are highly resistant to aromatic petroleum and chlorinated solvents, acids, alkalies, grease, oils, and Freon®. Nitrile formulation is free of inert fillers and silicone for superior chemical resistance. Gloves contain no natural rubber proteins and will not cause allergic reactions. Also resist punctures, snags, cuts, and abrasions.
Color: Green
Certifications: In compliance with 21 CFR 170-199. CE marked.
Caution: Note: Not recommended for use with ketones, strong oxidizing acids, and organic chemicals containing nitrogen.
2 Options Available Below
Product Details & Documents
Offer hand protection for industrial applications such as plating, metal finishing, chemical processing, abrasive material handling, maintenance, and acid etching.
- Curved fingers and tapered wrist for natural, comfortable wear
- Easy donning and doffing
- Flock lining absorbs perspiration for added comfort
- Flat cuffs
- Hand-specific
Double-dipped gloves are highly resistant to aromatic petroleum and chlorinated solvents, acids, alkalies, grease, oils, and Freon®. Nitrile formulation is free of inert fillers and silicone for superior chemical resistance. Gloves contain no natural rubber proteins and will not cause allergic reactions. Also resist punctures, snags, cuts, and abrasions.
Color: Green
Certifications: In compliance with 21 CFR 170-199. CE marked.
Caution: Note: Not recommended for use with ketones, strong oxidizing acids, and organic chemicals containing nitrogen.
Specifications for Product Family
Specifications
- Catalog No:
- 32917-718
- 32930-112
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
Specifications
Product Family Options
Product Information
- ModelGlove sizeGlove ColorLengthThicknessAvailabilityPrice
- 32917-718Z-Pattern Grip TextureZ-Pattern Grip TextureAF-188Green33 cm (13")18 mil.Each (12UN)$1,066.28Paquete de 12$1,066.28Case de 6 (12UN)$5,646.51Supplier Number: 483428ZQKSpecifications:
More
Specifications
Cat. No.32917-718Ambidextrous / hand-specificHand-specificDisposable / reusableDisposable - 32930-112Diamond Grip TextureDiamond Grip TextureAF-49310Green39.4 cm (15½")22 mil.Paquete de 12$3,285.15Case de 4 (12UN)$3,580.81Supplier Number: 493420Specifications:
More
Specifications
Cat. No.32930-112Ambidextrous / hand-specificHand-specificDisposable / reusableDisposable