

Iron (II) chloride tetrahydrate ≥99% BAKER ANALYZED®
Catalog # JT2064-1
Supplier: Avantor
CAS Number:
MOC: Non-RegulatedCertificates
Specifications
- Pack type:Glass bottle
- Size:500 g
- Cat. no.:JT2064-1
- Supplier No.:2064-01
Specifications
Specification Test Results
- Assay (FeCl₂·4H₂O) (by KMnO₄ titrn):99.0 - 103.0 %
- Insoluble Matter:≤0.01 %
- Phosphate (PO₄):≤0.001 %
- Sulfate (SO₄):≤0.01 %
- Copper (Cu):≤0.005 %
- Zinc (Zn):≤0.005 %
- Trace Impurities - Arsenic (As):≤10 ppm
Specification Test Results
About this item
:
Melting Pt:677 °C
Density:1.926 g/cm³ (20 °C)
EINECS:231-843-4
ADR:8,III
:Management of Change (MOC) category = N
:198.81 g/mol
UN:3260
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:13478-10-9
Formula:FeCl₂·4H₂O
MDL Number:MFCD00149709