Order Entry
Mexico
ContactUsLinkComponent
Hydrochloric acid ( 0.5 mol) concentrated aqueous solution 0.1 N
Corrosive
Hydrochloric acid ( 0.5 mol) concentrated aqueous solution 0.1 N
Catalog # JT4655-1
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
Corrosive
Hydrochloric acid ( 0.5 mol) concentrated aqueous solution 0.1 N
Catalog # JT4655-1
Supplier Number:  4655-01
CAS Number:  
MOC:  Non-RegulatedCertificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    LDPE ampoule, DILUT-IT®
  • Size:
    1 pack
  • Cat. no.:
    JT4655-1
  • Supplier No.:
    4655-01

Specifications

Specification Test Results

  • Normality (by titrimetry):
    Passes Test

Specification Test Results

About this item

 :
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,II
 :Management of Change (MOC) category = N
 :36.46 g/mol
 :GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324