Order Entry
Mexico
ContactUsLinkComponent
Hexane (mixture of isomers) ≥95% GC2® for gas chromatography, for capillary GC analysis
Flammable
Health
Irritant
Environment
Hexane (mixture of isomers) ≥95% GC2® for gas chromatography, for capillary GC analysis
Catalog # BJGC215-4
Product available on GSA Advantage®
CAS Number:  
undefined
Flammable
Health
Irritant
Environment
Hexane (mixture of isomers) ≥95% GC2® for gas chromatography, for capillary GC analysis
Catalog # BJGC215-4
Supplier Number:  GC215-4
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    4 L
  • Cat. no.:
    BJGC215-4
  • Supplier No.:
    GC215-4

Specifications

Specification Test Results

  • Electron Capture GC (as lindane):
    2 ppt max.
  • Flame Ionization GC (as nonane):
    1 ppb max.
  • GC Analysis (n-Hexane):
    95 % min.
  • GC Analysis (total C6 isomers):
    99.9 % min.
  • Residue:
    1 mg/L max.
  • Water by Karl Fischer Titration:
    0.01 % max.

Specification Test Results

About this item

 :
 :86.18 g/mol
Merck Index:13,04712
Boiling Pt:7.7 %(V)
Boiling Pt:68 - 70 °C (1013 hPa)
Melting Pt:–95 °C
Melting Pt:-95 °C
Density:0.66 g/cm³ (20 °C)
Flash Pt:–26 °C
Flash Pt:-22 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD02179311