Order Entry
Mexico
ContactUsLinkComponent
EDTA (Ethylenediamine tetraacetic acid) 99.4-100.6%, powder BAKER ANALYZED® ACS analytical reagent
Irritant
EDTA (Ethylenediamine tetraacetic acid) 99.4-100.6%, powder BAKER ANALYZED® ACS analytical reagent
Catalog # JT8991-1
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
Irritant
EDTA (Ethylenediamine tetraacetic acid) 99.4-100.6%, powder BAKER ANALYZED® ACS analytical reagent
Catalog # JT8991-1
Supplier Number:  8991-01
CAS Number:  
MOC:  Non-RegulatedCertificates

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Size:
    500 g
  • Cat. no.:
    JT8991-1
  • Supplier No.:
    8991-01

Specifications

Specification Test Results

  • ACS - Assay (C₁₀H₁₆N₂O₈):
    99.4 - 100.6 %
  • ACS - Insoluble in Dilute NH₄OH:
    ≤0.005 %
  • ACS - Residue after Ignition:
    ≤0.2 %
  • ACS - Nitrilotriacetic Acid:
    ≤0.1 %
  • ACS - Calcium (Ca):
    ≤0.001 %
  • ACS - Heavy Metals (as Pb):
    ≤0.001 %
  • ACS - Iron (Fe):
    ≤0.005 %
  • Trace Impurities - Magnesium (Mg):
    ≤5 ppm

Specification Test Results

About this item

 :
 :292.25 g/mol
Merck Index:13,03546
Boiling Pt:614 °C (1013 hPa)
Boiling Pt:614 °C (1013 hPa)
Melting Pt:220 °C
Density:0.86 g/cm³ (20 °C)
 :Management of Change (MOC) category = N
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:200-449-4
REACH:01-2119486399-18
 :GHS07
CAS Number:60-00-4
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541
Flash Pt:≥ 100 °C