Order Entry
Mexico
ContactUsLinkComponent
Sodium formate ≥99% BAKER ANALYZED® ACS
Sodium formate ≥99% BAKER ANALYZED® ACS
Catalog # JT3700-5
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
Sodium formate ≥99% BAKER ANALYZED® ACS
Catalog # JT3700-5
Supplier Number:  3700-05
CAS Number:  
MOC:  Non-RegulatedCertificates

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Size:
    2.5 kg
  • Cat. no.:
    JT3700-5
  • Supplier No.:
    3700-05

Specifications

Specification Test Results

  • ACS - Assay (HCOONa):
    ≥99.0 %
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Chloride (Cl):
    ≤0.001 %
  • ACS - Sulfate (SO₄):
    ≤0.001 %
  • ACS - Calcium (Ca):
    ≤0.005 %
  • Trace Impurities - Iron (Fe):
    ≤5 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm

Specification Test Results

About this item

 :
 :68.01 g/mol
Merck Index:13,08694
Melting Pt:257.31 °C
Density:1.919 g/cm³ (20 °C)
 :Management of Change (MOC) category = N
Storage Temperature:Ambient
EINECS:205-488-0
REACH:01-2119486468-21
CAS Number:141-53-7
Formula:HCO₂Na
MDL Number:MFCD00013101