Order Entry
Mexico
ContactUsLinkComponent
EDTA disodium salt dihydrate 0.1 M analytical reagent
EDTA disodium salt dihydrate 0.1 M analytical reagent
Catalog # RC273032
Supplier:  RICCA CHEMICAL CO
Small Business Enterprise
CAS Number:  
undefined
EDTA disodium salt dihydrate 0.1 M analytical reagent
Catalog # RC273032
Supplier:  RICCA CHEMICAL CO
Supplier Number:  R2730000-1A
Small Business Enterprise
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Size:
    32 oz.
  • Cat. no.:
    RC273032
  • Supplier No.:
    R2730000-1A

Specifications

Specification Test Results

  • Appearance:
    Clear, colorless, odorless
  • Assay at 20 °C (traceable to NIST SRM 915):
    0.1000 ± 0.0001 M at 20.0 °C

Specification Test Results

About this item

 :
 :372.24 g/mol
Merck Index:13,03543
Melting Pt:255 °C
Storage Temperature:Ambient
EINECS:205-358-3
CAS Number:6381-92-6
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037