Water ACS, Reagent Grade analytical reagent ASTM Type I, ASTM Type II Packaged in glass containers
Glass container, 1 gal.
Supplier: RICCA CHEMICAL CO
Número de proveedor #: R9151000-4C
Water ACS, Reagent Grade analytical reagent ASTM Type I, ASTM Type II Packaged in glass containers
Catalog # RC91511
Supplier: RICCA CHEMICAL CO
Small Business Enterprise
CAS Number: 7732-18-5
Jump to:
Product Details & Documents
Nonsterile. Deionized, double distilled quality. Meets specifications for ASTM. Type II water.
Synonyms: Hydrogen oxide
- CAS Number:7732-18-5
- Formula:H₂O
- MDL Number:MFCD00011332
- Molecular Weight:18.02 g/mol
- Merck Index:13,10098
- Boiling Pt:100 °C (1013 hPa)
- Boiling Pt:100 °C
- Melting Pt:0 °C
- Density:1 g/cm³ (4 °C)
- Storage Temperature:Ambient
- EINECS:231-791-2
Documents
Specifications
Specification Test Results
- Appearance:Colorless liquid
- Chloride (Cl):max 1 ppb
- Conductivity at 25°C:max 0.056 µS/cm
- Heavy Metals (as Pb):max 0.01 ppm
- Nitrate (NO₃):max 0.4 ppm
- Organic Carbon (TOC):max 50 ppb
- Oxidizable Substances (Permanganate Retention):To Pass Test
- Phosphate (PO₄):max 1.0 ppm
- Silicate (as SiO₂):max 3 ppb
- Sodium (Na):max 1 ppb
- Sulfate (SO₄):max 1.0 ppm
Specifications
- Pack type:Glass container
- Size:1 gal.
- Cat. No.:RC91511
- Environmental certification:EPA safer chemical solvent (Green circle rating); Chem21 solvent selection guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



