Water analytical reagent
Supplier: RICCA CHEMICAL CO
Certificates
About this item
CAS Number:7732-18-5
:
Formula:H₂O
MDL Number:MFCD00011332
:18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Storage Temperature:Ambient
EINECS:231-791-2
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer chemical solvent (green circle rating); Chem21 solvent selection guide recommended by default and after discussion
Specifications
Specification Test Results
- Appearance:Colorless liquid
- Chloride (Cl):max 1 ppb
- Conductivity at 25°C (Method: SQCP031):max 0.056 µS/cm
- Heavy Metals (as Pb):max 0.01 ppm
- Nitrate (NO₃):max 0.4 ppm
- Organic Carbon (TOC):max 50 ppb
- Oxidizable Substances (Permanganate Retention):To Pass Test
- Phosphate (PO₄):max 1.0 ppm
- Silicate (as SiO₂):max 3 ppb
- Sodium (Na):max 1 ppb
- Sulfate (SO₄):max 1.0 ppm
Specification Test Results
Related Information
Certificates
Search by Lot NumberRC91505
Images 360
Ricca Chemicals High Purity Water
Product Literature
Frequently Bought Together