Water ACS for HPLC
Catalog # BJAH365-4
Supplier: HONEYWELL INTERNATIONAL INC MX
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:4 L
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer chemical solvent (green circle rating); Chem21 solvent selection guide recommended by default and after discussion
- Cat. no.:BJAH365-4
- Supplier No.:AH365-4
Specifications
Specification Test Results
- HPLC Gradient @ 200 nm:0.005 AU max.
- UV Absorbance @ 250 nm:0.005 AU max.
- Microbial Contamination:Not detected
- Refractive Index (20ºC):1.3320 - 1.3340
- Residue:1 mg/L max.
- Specific Conductance (25ºC):2 Micromhos max.
- Substances Reducing Permanganate:Passes test
- Total Organic Carbon:50 ppb max.
- UV Absorbance @ 190 nm:0.01 AU max.
- UV Absorbance @ 200 nm:0.01 AU max.
Specification Test Results
About this item
CAS Number:7732-18-5
:
Formula:H₂O
MDL Number:MFCD00011332
:18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Storage Temperature:Ambient
EINECS:231-791-2