Order Entry
Mexico
Orders LinkContactUsLinkComponent
Water, high purity

Water, high purity

Plastic container, 1 gal.
Catalog # 72060-092
Número de proveedor #:  9801
Water, high purity
Catalog # 72060-092
CAS Number:  7732-18-5
Water, high purity

Plastic container, 1 gal.

CAS Number:  7732-18-5
Price
$3,825.96
Log in for availability
Case (4UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Ideal for reconstituting chemistry or coagulation products, preparing analytical standards, or rinsing delicate electrodes. Prepared at 18 megohm/cm specific resistance using reverse osmosis, mixed bed deionization, activated carbon filtration, and final filtration to 0.2µm. Contains no preservatives and is essentially free of soluble organics and inorganic particulate contaminants. Certificate of Analysis is available upon request.

Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2

Documents

Specifications

Specifications

  • Pack type:
    Plastic container
  • Size:
    1 gal.

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.