Order Entry
Mexico
ContactUsLinkComponent
EDTA disodium salt dihydrate ≥99%, crystals BAKER ANALYZED® ACS
EDTA disodium salt dihydrate ≥99%, crystals BAKER ANALYZED® ACS
Catalog # JT8993-1
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
EDTA disodium salt dihydrate ≥99%, crystals BAKER ANALYZED® ACS
Catalog # JT8993-1
Supplier Number:  8993-01
CAS Number:  
MOC:  Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Size:
    500 g
  • Cat. no.:
    JT8993-1
  • Supplier No.:
    8993-01

Specifications

Specification Test Results

  • Assay (C₁₀H₁₄N₂Na₂O₈·2H₂O):
    99.0 - 101.0 %
  • Insoluble Matter:
    ≤0.005 %
  • pH of 5% Solution at 25°C:
    4.0 - 6.0
  • Nitrilotriacetic Acid (by polarography):
    ≤0.1 %
  • Heavy Metals (as Pb):
    ≤0.005 %
  • Iron (Fe):
    ≤0.01 %

Specification Test Results

About this item

CAS Number:6381-92-6
 :
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037
 :372.24 g/mol
Merck Index:13,03543
Melting Pt:255 °C
 :Management of Change (MOC) category = N
Storage Temperature:Ambient
EINECS:205-358-3