

Diethyl ether ≥99.9% ACS for HPLC, for spectrophotometry
Catalog # BJ107-4
Supplier: HONEYWELL INTERNATIONAL INC MX
Product available on GSA Advantage®
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:4 L
- Cat. no.:BJ107-4
- Supplier No.:107-4
Specifications
Specification Test Results
- Electron Capture GC:10 ng/L max.
- Water by Karl Fischer Titration:0.03 % max.
- GC Analysis:99.9 % min.
- Peroxides (as H₂O₂) at time of packaging:1 ppm max.
- Refractive Index (20ºC):1.3516 - 1.3526
- Residue:1 mg/L max.
- UV Absorbance @ 250 nm:0.08 AU max.
- UV Absorbance @ 275 nm:0.01 AU max.
- UV Absorbance @ 300 nm:0.005 AU max.
- UV Cutoff:215 nm max.
Specification Test Results
About this item
:
:74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–117.4 °C
Melting Pt:-123 °C
Density:0.7135 g/cm³ (20 °C)
Flash Pt:–45 °C
Flash Pt:-40 °C
Storage Temperature:Ambient
EINECS:200-467-2
UN:1155
ADR:3,I
REACH:01-2119535785-29
:GHS02
:GHS07
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646