
Ethanol absolute, anhydrous KOPTEC USP, pure 200 Proof
Catalog # 71002-426
Supplier: DECON LABS
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:1 gal.
- Cat. no.:71002-426
- Duty Status:Duty paid
- Supplier No.:V1001GTP
Specifications
Specification Test Results
- Max. UV Absorbance at 240 nm:0.40 AU
- Max. UV Absorbance at 250 nm:0.3 AU
- Max. UV Absorbance at 260 nm:0.3 AU
- Max. UV Absorbance at 270 nm:0.1
- Acidity:30 ppm max.
- Alkalinity:Pass test
- Assay:200 proof (99.5% min.)
- Benzene:2 ppm max.
- Clarity:Pass
- Color:Pass test
- Methanol:100 ppm max.
- Non-Volatile Residue:25 ppm max.
- Relative Density at 20°C (BP/EP):0.790–0.793
- Specific Gravity at 15.0°C (JP):0.797 max.
- Specific Gravity at 15.56°C (USP):0.7962 max.
- Sum of Acetal & Acetaldehyde:10 ppm max.
- Water:0.1% (v/v) max.
Specification Test Results
About this item
:
:46.07 g/mol
Merck Index:12,03806
Boiling Pt:78.3 °C (1013 hPa)
Boiling Pt:78.3 °C (1013 hPa)
Melting Pt:–117 °C
Melting Pt:-117 °C
Density:0.7895 g/cm³ (20 °C)
Flash Pt:12 °C
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
REACH:01-2119457610-43
:GHS02
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568