Order Entry
Mexico
ContactUsLinkComponent
1,2-Dichloroethane ≥99.0% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
1,2-Dichloroethane ≥99.0% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
Catalog # MK496604
Supplier:  Avantor
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
1,2-Dichloroethane ≥99.0% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
Catalog # MK496604
Supplier:  Avantor
CAS Number:  
MOC:  Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Size:
    500 ml
  • Environmentally Preferable:
  • Cat. no.:
    MK496604
  • Supplier No.:
    4966-04

Specifications

Specification Test Results

  • ACS - Assay (C₂H₄Cl₂) (by GC, corrected for water):
    ≥99.0 %
  • Appearance:
    Passes Test
  • ACS - Color (APHA):
    ≤10
  • ACS - Residue after Evaporation:
    ≤0.002 %
  • ACS - Titrable Acid (meq/g):
    ≤0.0003
  • ACS - Water (H₂O)(by KF, coulometric):
    ≤0.03 %

Specification Test Results

About this item

Synonyms: Ethylene chloride, Ethylene dichloride, Dichloroethane
 :
 :98.96 g/mol
Merck Index:13,03831
Boiling Pt:83.5 °C (1013 hPa)
Boiling Pt:83.5 °C (1013 hPa)
Melting Pt:–35.5 °C
Melting Pt:-35.5 °C
Density:1.254 g/cm³ (20 °C)
Flash Pt:13 °C
Flash Pt:13 °C
Storage Temperature:Ambient
EINECS:203-458-1
UN:1184
ADR:3,II
 :Management of Change (MOC) category = N
 :GHS02
 :GHS08
 :GHS07
CAS Number:107-06-2
Formula:ClH₂CCH₂Cl
MDL Number:MFCD00000963