


N,N-Dimethylformamide ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
MOC: Non-Regulated
About this item
CAS Number:68-12-2
:
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284
:73.09 g/mol
Merck Index:13,03269
Boiling Pt:153 °C (1013 hPa)
Melting Pt:-61 °C
Density:0.949 g/cm³ (20 °C)
Flash Pt:57.5 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
REACH:01-2119475605-32
:GHS02
:GHS08
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:Clear, colorless liquid (can be slightly yellow)
- Water (K.F.):Less than 0.15%
- Residue:Less than 0.005% (0.0010 g/21 mL)
- GC Purity:Greater than 99.8%
- Color (APHA):Less than 10
- Titrable acid:Less than 0.5 meq/g (Not more than 1.0 mL of 0.01N sodium hydroxide)
- Titrable base:Less than 3 meq/g (Not more than 3.0 mL of 0.01N sodium hydroxide)
Specification Test Results
Related Information
Certificates
Search by Lot Number12777752.pdf BDH1117-19L_BSETSE BDH1117-19L_P BDH1117-204L_BSETSE BDH1117-4LG_BSETSE
BSE/TSE Certificates
More
19-20 l Close Top Steel Can with Handle
Packaging Information
Avantor Portfolio of Chemicals Avantor Top Rated Guide
Product Literature
spec_BDH1117
Product specification
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together