AK Natural Latex Cleanroom Gloves, Honeywell Safety
For cleanroom hand tasks and protection from acids and caustics.
- Natural rubber latex, powder free, case hardened
- Outstanding puncture and tear resistance
- Reduced ionic and extractable contamination
- Positive grip in wet/dry applications
- Hand specific, rolled cuff, diamond-embossed finish
These natural rubber latex gloves are powder-free and case-hardened with outstanding puncture and tear resistance. Reduced ionic and extractable contamination. Positive grip in wet/dry applications. Size embossed instead of inked to eliminate ink contamination. Hand-specific. Rolled cuff. Diamond embossed finish. Color: orange.
: Meet IES RP-005.2 (modified) particulate and extractables test. Meet ASTM 739 permeation standard.
- Catalog No:
- 32888-210
- 32888-206
- 32888-208
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Latex / latex-free:
- Latex
- Latex
- Latex
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
Frequently Bought Together