Order Entry
Mexico
ContactUsLinkComponent
Citric acid monohydrate 99.5-100.5% BP|Ph. Eur.|USP|E330
Irritant
Citric acid monohydrate 99.5-100.5% BP|Ph. Eur.|USP|E330
Catalog # EM1.00242.5000
CAS Number:  
undefined
Irritant
Citric acid monohydrate 99.5-100.5% BP|Ph. Eur.|USP|E330
Catalog # EM1.00242.5000
Supplier Number:  1.00242.5000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    5 kg
  • Cat. no.:
    EM1.00242.5000
  • Supplier No.:
    1.00242.5000

Specifications

Specification Test Results

  • Assay (alkalimetric, calculated on anhydrous substance):
    99.5 - 100.5 %
  • Identity (IR-spectrum):
    passes test
  • Appearance:
    white to almost white or colorless crystals
  • Appearance of solution (200 g/l; water):
    clear (≤ 3 NTU) and not more intense in color than reference solution Y₇, BY₇ or GY₇
  • Sulfate (SO₄):
    ≤ 50 ppm
  • Heavy metals (as Pb):
    ≤ 5 ppm
  • Al (Aluminium):
    ≤ 0.2 ppm
  • As (Arsenic):
    ≤ 1 ppm
  • Hg (Mercury):
    ≤ 1 ppm
  • Pb (Lead):
    ≤ 0.5 ppm
  • Oxalates (as C₂H₂O₄):
    ≤ 100 ppm
  • Residual solvents (ICH (Q3C)):
    excluded by manufacturing process
  • Readily carbonisable substance:
    passes test
  • Sulfated ash (600 °C):
    ≤ 0.05 %
  • Water (according to Karl Fischer):
    7.5 - 8.8 %
  • Bacterial endotoxins:
    0.5

Specification Test Results

About this item

 :
 :210.14 g/mol
Merck Index:13,02350
Melting Pt:135…152 °C
Density:1.552 g/cm³ (20 °C)
Flash Pt:173.9 °C (closed cup)
EINECS:201-069-1
 :GHS07
Storage Temperature:Ambient
CAS Number:5949-29-1
Formula:C₆H₈O₇.H₂O
MDL Number:MFCD00149972
Melting Pt:135 - 152 °C