Order Entry
Mexico
ContactUsLinkComponent
 
Chloroform ≥99.9% stabilised ACS for HPLC, for spectrophotometry
Catalog # BJ049-4
Product available on GSA Advantage®
CAS Number:  
undefined
Chloroform ≥99.9% stabilised ACS for HPLC, for spectrophotometry
Catalog # BJ049-4
Supplier Number:  049-4
Product available on GSA Advantage®
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass bottle
  • Size:
    4 L
  • Cat. no.:
    BJ049-4
  • Supplier No.:
    049-4

Specifications

Specification Test Results

  • Acidity (as HCl):
    1 mg/L max.
  • Chloride:
    10 mg/L max.
  • Electron Capture GC:
    10 ng/L max.
  • GC Analysis:
    99.9 % min.
  • Refractive Index (20ºC):
    1.4454 - 1.4464
  • Residue:
    1 mg/L max.
  • Substances Darkened by Sulfuric Acid:
    Passes test
  • UV Cutoff:
    245 nm max.
  • Water by Karl Fischer Titration:
    0.02 % max.

Specification Test Results

About this item

 :
 :119.38 g/mol
Merck Index:13,02160
Boiling Pt:62 °C
Boiling Pt:61.7 °C (1013 hPa)
Melting Pt:–64 °C
Melting Pt:-63 °C
Density:1.4832 g/cm³ (20 °C)
Flash Pt: Does not flash
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
 :Stabilised with amylene
REACH:05-2114614906-45
 :GHS06
 :GHS08
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826