Acetonitrile ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Catalog # EM-AX0156-1
Supplier: THOMAS SCIENTIFIC INC MX
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Glass bottle
- Size:4 L
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent selection guide recommended by default
- Cat. no.:EM-AX0156-1
- Supplier No.:AX0156-1
Specifications
Specification Test Results
- Assay (GC):99.9% min
- Color (APHA):10APHA max
- Filtered through 0.2 µm filter:To pass test
- Fluorescence as quinine at 254 nm:1ppb max
- Fluorescence as quinine at 365 nm:1ppb max
- Form:Clear liquid
- Gradient at 210 nm:2mAU max
- Gradient at 254 nm:0.3mAU max
- Identity (IR-spectrum):Conforms
- Metals, suitable for LC/MS:To pass test
- MS purity, ESI(+), largest response as reserpine:50ppb max
- MS purity, ESI(-), largest response, p-nitrophenol:50ppb max
- Residue after evaporation:1ppm max
- Titrable acid:1µeq/g max
- Titrable base:0.5µeq/g max
- UV Abs. at 200 nm:0.1AU max
- UV Abs. at 210 nm:0.04AU max
- UV Abs. at 220 nm:0.02AU max
- UV Abs. at 230 nm:0.01AU max
- UV Abs. at 240 nm:0.01AU max
- UV Abs. at 250 nm:0.01AU max
- UV Abs. at 270 nm:0.01AU max
- UV Abs. at 400-280 nm:0.005AU max
- UV Cut-off:190nm max
- Water (H2O):0.02% max
Specification Test Results
About this item
Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
:
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:81.6 °C (1013 hPa)
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45.7 °C
Melting Pt:-45.7 °C
Density:0.782 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
:GHS02
:GHS07
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878