

Acetonitrile ≥99.9% ACS for HPLC, for spectrophotometry
Catalog # BJ018-4
Supplier: HONEYWELL INTERNATIONAL INC MX
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:4 L
- Cat. no.:BJ018-4
- Supplier No.:018-4
Specifications
Specification Test Results
- Carbonyl Compounds (as HCHO):10 ppb max.
- Water by Karl Fischer Titration:0.01 % max.
- GC Analysis:99.9 % min.
- HPLC Gradient @ 215 nm:0.005 AU max.
- HPLC Gradient @ 254 nm:0.001 AU max.
- Refractive Index (20ºC):1.3434 - 1.3446
- Residue:1 mg/L max.
- UV Absorbance @ 200 nm:0.05 AU max.
- UV Absorbance @ 210 nm:0.04 AU max.
- UV Cutoff:190 nm max.
Specification Test Results
About this item
:
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:82 °C (1.013 hPa)
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–46 °C
Melting Pt:-45.7 °C
Density:0.782 g/cm³ (20 °C)
Flash Pt:6 °C
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
:GHS02
:GHS07
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878