VWR® Traceable® Four-Channel Alarm Timer with Clock
Product Details & Documents
Microcomputer chip permits setting four separate channels for simultaneous countup or countdown procedures.
- Massive ³/₄" high display allows viewing from across the room
- Extra-loud, high decibel alarms for 1 minute and may be silenced manually
- Timing capacity to 99 hours, 59 minutes and 59 seconds with a 1 second resolution
- Remarkable memory returns display to previously programmed countdown time at the touch of a button
Large 1.9 cm LCD permits viewing from across the lab. Additional features include stopwatch, time-out, and time-of-day display. Use all four channels simultaneously as count-up stopwatches or in any combination of countup/countdown. The touch of a button returns the display to the previously programmed countdown time. Extra-loud, high-decibel alarm sounds for one minute or may be silenced manually. Finger-size keys make it easy to set times and change channels. Magnetic back allows timer to be placed on any metal surface. Unit also has a flip-open easel for standing on the lab bench, a spring fastener for clipping to a notebook/clipboard, and a hole for use with a lanyard.
When zero is reached the timer starts counting up to show time elapsed since alarming.
Ordering information: Supplied with a one-year, 1.5 V silver-oxide battery. Replacement battery is available separately.
Delivery information: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Documents
Certificates
Search by Lot NumberAvantor Top Rated Guide
Product Literature
VWR Traceable Timers
Promotion Material
Specifications
- W×H×D:7×6.4×1.3 cm (2¾×2½×½")