Acetic acid glacial ≥99.7% (by GC, corrected for water content), BAKER ANALYZED® A.C.S. Reagent ACS Reagent aldehyde-free, J.T.Baker®
About this item
The quality of BAKER ANALYZED™ ACS Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These acids are tested for common trace metals.
: IMPORTANT: Material will freeze if stored below 17 °C (63 °F).
- CAS Number:64-19-7
- Formula:H₃CCOOH
- MDL Number:MFCD00036152
- Molecular Weight:60.05 g/mol
- Boiling Pt:118 °C (1013 hPa)
- Melting Pt:17 °C
- Density:1.05 g/cm³ (20 °C)
- Flash Pt:38.5 °C
- EINECS:200-580-7
- ADR:8,II
- :Management of Change (MOC) category = N
- Storage Temperature:Ambient
- UN:2789
- REACH:01-2119475328-30
2 Options Available Below
Product Details & Documents
The quality of BAKER ANALYZED™ ACS Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These acids are tested for common trace metals.
: IMPORTANT: Material will freeze if stored below 17 °C (63 °F).
- CAS Number:64-19-7
- Formula:H₃CCOOH
- MDL Number:MFCD00036152
- Molecular Weight:60.05 g/mol
- Boiling Pt:118 °C (1013 hPa)
- Melting Pt:17 °C
- Density:1.05 g/cm³ (20 °C)
- Flash Pt:38.5 °C
- EINECS:200-580-7
- ADR:8,II
- :Management of Change (MOC) category = N
- Storage Temperature:Ambient
- UN:2789
- REACH:01-2119475328-30
Specifications for Product Family
Specification Test Results
- Assay (by GC, corrected for water):≥99.7 %
- Color (APHA):≤10
- Dilution Test:Passes Test
- Residue after Evaporation:≤0.001 %
- Acetic Anhydride ((CH₃CO)₂O):≤0.01 %
- Chloride (Cl):≤1 ppm
- Sulfate (SO₄):≤1 ppm
- Trace Impurities - Iron (Fe):≤0.2 ppm
- Substances Reducing Dichromate:Passes Test
- Substances Reducing Permanganate:Passes Test
- Titrable Base (meq/g):≤0.0004
- Acetaldehyde:≤0.005 %
- Sensitivity:Passes Test
- Trace Impurities - Arsenic and Antimony (as As):≤50.0 ppb
- Trace Impurities - Boron (B):≤100.0 ppb
- Trace Impurities - Chromium (Cr):≤200.0 ppb
- Trace Impurities - Cobalt (Co):≤100.0 ppb
- Trace Impurities - Copper (Cu):≤100.0 ppb
- Trace Impurities - Heavy Metals:≤0.5 ppm
- Trace Impurities - Lead (Pb):≤300.0 ppb
- Trace Impurities - Manganese (Mn):≤200.0 ppb
- Trace Impurities - Nickel (Ni):≤100.0 ppb
- Trace Impurities - Potassium (K):≤300.0 ppb
- Trace Impurities - Tin (Sn):≤100 ppb
- Trace Impurities - Titanium (Ti):≤300.0 ppb
- Trace Impurities - Zinc (Zn):≤200.0 ppb
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Glass bottle, clear in a single shipper500 mlEach(1/Itm/Bx)$852.79Case de 12 (1/ Itm/Bx)$10,233.44Supplier Number: 9508-02Specifications:MOC: Non-RegulatedMOC: Non-Regulated
- Glass bottle, clear in a single shipper2.5 LEach(1/Itm/Bx)$1,844.76Case de 6 (1/ Itm/Bx)$11,068.57Supplier Number: 9508-05Specifications:MOC: Non-RegulatedMOC: Non-Regulated
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



