Cryogenic Storage Boxes, Polycarbonate, Corning®
Product Details & Documents
These cryogenic storage boxes are suitable for storage down to –196°C (–320.8°F). These boxes feature a large marking spot and a clear polycarbonate lid that can be labeled to show vial contents. They are stackable and are available in two formats: 81-place boxes which will hold internally and externally threaded vials; and 100 count boxes, which will hold internally threaded vials.
- Reusable
- Ridges on the sides of the box make them easier to hold while wearing gloves
- Transparent lid with numeric grid
Designed for use with most cryogenic vials.
These boxes feature a large marking spot and a clear polycarbonate lid that can be labeled to show vial contents. They are stackable and are available in two formats: 81-place boxes which will hold internally and externally threaded vials; and 100 count boxes, which will hold internally threaded vials.
These cryogenic storage boxes are suitable for storage down to –196°C (–320.8°F).
Documents
Specifications
- No. of Holes:81
- Size:5/Pack, 10/Case
- For Vials:1 - 2 ml