Order Entry
Mexico
ContactUsLinkComponent
'Lid-Off' Lid Remover, Qorpak®
'Lid-Off' Lid Remover, Qorpak®
Catalog # 16199-000
Supplier:  QORPAK
Small Business Enterprise
undefined
'Lid-Off' Lid Remover, Qorpak®
Catalog # 16199-000
Supplier:  QORPAK
Supplier Number:  MET-08918
Small Business Enterprise
Specifications
  • Description:
    Lid Remover
  • Cat. no.:
    16199-000
  • Supplier No.:
    MET-08918
Specifications
About this item
To safely open tightly sealed pails, simply hook “Lid-Off” over the edge of the lid and lift.
Lid remains undamaged and can be opened and closed repeatedly.