VWR® Scoop, PTFE
80L x 30W x 20D mm (31/8x11/8x3/4")
Catalog # 89026-220
Supplier: VWR International
Jump to:
Product Details & Documents
Non-contaminating PTFE scoop useful for transferring frozen substances and other non-liquids.
- Chemically inert
- Non-contaminating
- PTFE construction
Scoop is suitable for use with frozen substances and other solids.
Specifications
- Dimensions:80L x 30W x 20D mm (31/8x11/8x3/4")