VWR® Spatula, PTFE-Coated
Catalog # 82027-534
Supplier: VWR International
Specifications
- Length:17.8 cm (7")
- Cat. no.:82027-534
Specifications
About this item
Stainless steel spatula features one rounded end and one flat end, with PTFE coating throughout.
- Stainless steel construction
- PTFE coating
- Nonsticking and non-corrosive
It can be used to transfer corrosive or sticky materials without scratching or damaging container surfaces.