Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® PTFE-Coated Forceps
VWR® PTFE-Coated Forceps
114 mm (4½")
Catalog # 82027-454
Supplier:   VWR International
VWR® PTFE-Coated Forceps
Catalog # 82027-454
Supplier:  VWR International
VWR® PTFE-Coated Forceps
VWR® PTFE-Coated Forceps
114 mm (4½")
Price
$353.10
Log in for availability
Each (1UN) Paquete de 12 (1UN) Case de 60 (1UN)
Product Details & Documents
Stainless steel forceps are PTFE-coated.
  • Stainless steel construction
  • Chemically resistant
  • Nonsticking and non-corroding
Forceps are coated with PTFE to imbue forceps with heat and chemical resistance.
Documents
Specifications
  • Length:
    114 mm (4½")