Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Self-Closing Forceps
VWR® Self-Closing Forceps
127 mm (5")
Catalog # 82027-452
Supplier:   VWR International
VWR® Self-Closing Forceps
Catalog # 82027-452
Supplier:  VWR International
VWR® Self-Closing Forceps
VWR® Self-Closing Forceps
127 mm (5")
Price
$103.10
Log in for availability
Each (1UN) Paquete de 12 (1UN) Case de 60 (1UN)
Product Details & Documents
Stainless steel self-closing forceps feature straight, fine tips and a smooth grip.
  • Stainless steel construction
  • Fine tips
  • Self-closing design
Forceps are passivated and autoclavable.
Specifications
  • Length:
    127 mm (5")