Nalgene® Filter Forceps, Thermo Scientific
Straight Tip
28198-976
: DS0399-0002
Stainless steel blades have serrated grips for handling membrane filters without risk of damage or contamination
Available in bent tip and straight tip. Bent tip forceps feature X-action design that allows membrane to be held without applying constant pressure. Autoclavable.
- Description:Straight Tip