Order Entry
Mexico
ContactUsLinkComponent
Dissecting Forceps, Fine Tip, Serrated, Grobet USA
Dissecting Forceps, Fine Tip, Serrated, Grobet USA
Catálogo # 25718-012
undefined
Dissecting Forceps, Fine Tip, Serrated, Grobet USA
Catálogo # 25718-012
Supplier Number:  57.929
Specifications
  • Length:
    117 mm (4⅝")
  • Cat. no.:
    25718-012
  • Supplier No.:
    57.929
Specifications
About this item
Made of nickel-plated, polished steel with fine, straight points having fine file-cut serrations on inner side
Provided with guide pin.