Order Entry
Mexico
ContactUsLinkComponent
VWR® Beaker Tongs, Fiberglass Coating
Catalog # 82027-374
Supplier:  VWR International
undefined
VWR® Beaker Tongs, Fiberglass Coating
Catalog # 82027-374
Supplier:  VWR International
Specifications
  • Length:
    31.8 cm (12½")
  • Cat. no.:
    82027-374
Specifications
About this item
Tongs feature a stainless steel construction, a riveted joint, and fiberglass coating for use in high-temperature applications.
  • Stainless steel construction
  • For use in high-temperature applications
Designed for the safe and simple handling of beakers and similar vessels from 50 to 2000 mL capacities.