Order Entry
Mexico
ContactUsLinkComponent
SP Bel-Art Sterileware® Sterile Mini Tongs, Bel-Art Products, a part of SP
SP Bel-Art Sterileware® Sterile Mini Tongs, Bel-Art Products, a part of SP
Catalog # 47733-014
undefined
SP Bel-Art Sterileware® Sterile Mini Tongs, Bel-Art Products, a part of SP
Catalog # 47733-014
Supplier Number:  F37944-0000
Specifications
  • Length:
    10.8 cm (4¼")
  • Cat. no.:
    47733-014
  • Supplier No.:
    379440000
Specifications
About this item
These small, lightweight tongs are ideal for sampling small quantities of material ranging from food to pharmaceuticals.
  • Lightweight
  • One-piece polystyrene plastic construction
  • Ridged jaws securely hold wet or dry samples
  • Individually packaged
  • Sterilized via gamma irradiation
Their ridged jaws securely hold wet or dry samples
Tongs are individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilization.
One-piece, sterile tongs are made from polystyrene plastic.
Ordering information: Individually wrapped and packed for one-time use and disposal.